Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Y-Connector, Stainless Steel, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
Y-3 EA Y-8 EA Y-1 EA Y0-6SS Y0-2.5SS Y0-3SS Y0-5SS
MFLX30620-24EA 92.6 USD InStock
MFLX30620-24 MFLX30620-23 MFLX30620-22 MFLX30620-21 MFLX31209-55 MFLX31209-57 MFLX31209-56
Masterflex® Union Fittings, Hose Barb, Y-Connector, Stainless Steel, Avantor®
Fittings and Connectors
Ensure a tight seal every time.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty