Would you like to log in again?
Union Fittings, Male Threaded Adapter Nipple, Straight
Supplier: VWR International
N12PP(5 PACK) N2NK7-MF 10PK N6HDPE(100 PACK) N4PP(10 PACK) 40615-24 N16PP(5 PACK) N12NN(6 PACK) N6PP(10 PACK) N2PP(100/PK) N2NN(6 PACK) N8-8 N6HDPE(10 PACK) N4HDPE(10 PACK) N6-6 N2HDPE(10 PACK) N4-4 N4NN(10 PACK) N8HDPE(10 PACK) N2NN(60 PACK) N12-12 N2HDPE(100 PACK) N8NK7-MF 10PK N6NN(6 PACK) N2PP(10 PACK) MAKE-UP N12NK7-MF 10PK N8PP(10 PACK) N8NN(10 PACK) N16NK7-MF 10PK N6NK7-MF 10PK N12HDPE(10 PACK) N2-2
MFLX30627-24EA 102 USD InStock
MFLX30627-24 MFLX30627-25 MFLX30623-20 MFLX30623-21 MFLX30629-02 MFLX30627-28 MFLX30623-24 MFLX30623-25 MFLX30623-26 MFLX30623-27 MFLX30623-28 MFLX30623-29 MFLX40610-92 MFLX40615-23 MFLX40610-93 MFLX40615-24 MFLX40610-90 MFLX40610-91 MFLX40615-25 MFLX40610-94 MFLX40610-95 MFLX40615-26 MFLX40615-27 MFLX40615-28 MFLX06546-31 MFLX30623-31 MFLX06349-71 MFLX06546-29 MFLX06546-24 MFLX06546-22 MFLX06546-26 MFLX40640-90
Union Fittings, Male Threaded Adapter Nipple, Straight
Fittings and Connectors
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings provide a more permanent fluid path. Choose the size and material to meet your application needs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty