Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Tee, HDPE, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
T0-2-200HDPE (100 PACK) T0-4HDPE(100 PACK) T0-12HDPE(10 PACK) T0-10HDPE-MF(10PK) T0-1.5-200HDPE(100 PACK) T0-3HDPE(10 PACK) T0-3HDPE-MF(10PK) T0-16HDPE-MF(10PK) T0-2-200HDPE-MF(10PK) T0-1-200HDPE (10 PACK) T0-6HDPE(10 PACK) T0-10HDPE(100 PACK) T0-1-200HDPE-MF(10PK) T0-5HDPE(100 PACK) T0-6HDPE(100 PACK) T0-8HDPE-MF(10PK) T0-5HDPE(10 PACK) T0-6HDPE-MF(10PK) T0-4HDPE(10 PACK) T0-12HDPE(100 PACK) T0-8HDPE(10 PACK) T0-2-200HDPE (10 PACK) T0-3HDPE(100 PACK) T0-12HDPE-MF(10PK) T0-1.5-200HDPE(10 PACK) T0-8HDPE(100 PACK) T0-4HDPE-MF(10PK) T0-10HDPE(10 PACK)
MFLX30623-60EA 29.7 USD InStock
MFLX30623-60 MFLX40623-64 MFLX30627-67 MFLX30623-67 MFLX90009-27 MFLX40613-01 MFLX30613-21 MFLX30613-25 MFLX40613-05 MFLX30627-60 MFLX30613-29 MFLX30613-09 MFLX30627-73 MFLX30623-71 MFLX30623-73 MFLX30627-79 MFLX30627-58 MFLX30627-77 MFLX40627-56 MFLX30623-77 MFLX40624-27 MFLX40623-56 MFLX30623-79 MFLX30623-58 MFLX30613-33 MFLX30613-13 MFLX30627-71 MFLX30613-17
Masterflex® Union Fittings, Hose Barb, Tee, HDPE, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty