Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Tee, Nylon, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
T0-2-200NN (10 PACK) T0-8NN-MF(10PK) T0-1-200NN (10 PACK) T0-1-200NN-MF(10PK) T0-2-200NN (100 PACK) T0-8NN(100 PACK) T0-10NN-MF(10PK) T0-12NN(10 PACK) T0-6NN(100 PACK) T0-8NN(10 PACK) T0-12NN-MF(10PK) CEA33-NR0 T0-2-200NN-MF(10PK) T0-1.5-200NN(100 PACK) T0-4NN-MF(10PK) T 112 5 PK T0-5NN(100 PACK) T0-3NN(100 PACK) T0-10NN(100 PACK) T0-3NN-MF(10PK) T0-16NN-MF(10PK) T0-16NN(10 PACK) T0-12NN(100 PACK) T0-6NN(10 PACK) T0-5NN(10 PACK) T0-4NN(100 PACK) T0-10NN(10 PACK) T0-6NN-MF(10PK) T0-3NN(10 PACK) T0-4NN(10 PACK) T 114 5 PK
MFLX30623-80EA 27.4 USD InStock
MFLX30623-80 MFLX40627-65 MFLX30623-61 MFLX30627-69 MFLX30623-63 MFLX40623-65 MFLX30623-69 MFLX31408-09 MFLX31408-08 MFLX30613-22 MFLX30613-26 MFLX40613-02 MFLX31500-30 MFLX30627-61 MFLX30627-80 MFLX40613-06 MFLX30627-75 MFLX40628-26 MFLX30623-72 MFLX30623-75 MFLX30627-78 MFLX30623-78 MFLX30627-59 MFLX30623-59 MFLX40624-29 MFLX30613-10 MFLX30613-30 MFLX30613-14 MFLX30613-34 MFLX30613-18 MFLX30627-72
Masterflex® Union Fittings, Hose Barb, Tee, Nylon, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty