Would you like to log in again?
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
FTLL007-6005 FTLL220-6005 (25PK) FTLL410-6005 (25PK) FTLL004-6005 (25PK) FTLL360-6005 (25PK) FTLLSB013-6005 (25PK) FTLL210-6005 (25PK) FTLL250-6005 (25PK) FTLLSB004-6005 (25PK) FTLL055-6005 (25PK) FTLL430-6005 (25PK) FTLL230-6005 (25PK) FTLL240-6005 (25PK) FTLL013-6005 (25PK) FTLL035-6005 (25PK)
MFLX30800-06EA 15.2 USD InStock
MFLX30800-06 MFLX30800-08 MFLX30800-02 MFLX30800-04 MFLX30800-10 MFLX45508-50 MFLX45508-00 MFLX45508-12 MFLX45508-01 MFLX45508-54 MFLX45508-16 MFLX45508-02 MFLX45508-03 MFLX45508-08 MFLX45508-06
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene, Avantor®
Fittings and Connectors
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty