Would you like to log in again?
CPC® Multi-Tube Quick-Disconnect Connectors
Supplier: CPC (Colder Products Company)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TMB1001 PTC16010 (25PK) TFB100112 TFB1001 SXF4202 10PK TMB1002 SXM42M3 SXF4201 PTC16020 (25PK) PTC2202096 (1/PK) SXF420112 PTC22020 (1/PK) TT1001 5PK PTC1602096 SXM1702 10PK PTC22010 (25PK) PTC16020 (1/PK) PTC22020 (25PK) SXM170112 SXFD4202 TFB1002 SXF420112 10PK TMB10M3 TFBD1002 SXM1702 TT1002 TMB1001 TT100212 SXFD42M3 SXM1701 TT1001 TFB1002 TT100212 5PK TMB1002 5PK PTC16010 (1/PK) TS10 TFBD1001 PTC22010 (1/PK) PTC2202096 (25PK) SXFD4201 TFBD1001 SXF4202 SXFD4201 TMB100112
MFLX31308-19EA 161 USD InStock
MFLX31308-19 MFLX31308-16 MFLX31308-17 MFLX31050-04BLK MFLX31308-14 MFLX31050-02BLK MFLX31307-86 MFLX31050-06BLK MFLX31051-27 MFLX31307-89 MFLX31307-88 MFLX31307-85 MFLX31307-88BLK MFLX31307-81 MFLX31307-75BLK MFLX31308-09BLK MFLX31307-96BLK MFLX31308-09 MFLX31308-07 MFLX31052-41 MFLX31050-01 MFLX31050-02 MFLX31308-06 MFLX31308-28 MFLX31050-01BLK MFLX31308-25 MFLX31052-24 MFLX31308-26 MFLX31050-05 MFLX31307-98 MFLX31050-06 MFLX31050-05BLK MFLX31308-24 MFLX31307-75 MFLX31050-03 MFLX31050-04 MFLX31307-93 MFLX31307-96 MFLX31307-73 MFLX31307-90 MFLX31307-91 MFLX31308-25BLK MFLX31307-93BLK MFLX31308-06BLK
CPC® Multi-Tube Quick-Disconnect Connectors
Fittings and Connectors
Save time—with one coupling you can connect two, six, or ten lines.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • One easy-to-use quick disconnect for multiple tubing lines
  • Multiline couplings offer quick transfers and minimize down time
These CPC® multi-tube connectors simplify the manual steps in your process for quick transfers and minimize your down time. These couplings make it easy to handle multiple tubing lines in your process that require connection and reconnection. They can disconnect multiple lines with the push of a button. Choose from the Twin Tube™, Sixtube™, and Tentube™ series couplings, providing two, six, or ten lines respectively.
The Twin Tube™ coupling provides one easy-to-use quick disconnect for two separate fluid lines. These non-valved couplings maintain two individual flow paths in one connection.
The Sixtube™ coupling provides one easy-to-use quick disconnect for up to six separate fluid lines. These are available with valves on either side for maximum design flexibility.
The Tentube™ coupling provides one easy-to-use quick disconnect for up to ten separate fluid lines. The These are available with valves on the body side in panel mount or in-line configurations for maximum design flexibility.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty