Would you like to log in again?
Masterflex® Luer Lock Fittings, Stopcock, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
RAC51-DF1 K-LL2W RAC36-DF1 K-LL2W K-LL3W RAC70-DF1
MFLX31200-90CS 0 USD InStock
MFLX31200-90 MFLX31200-92 MFLX31200-70 MFLX31200-91 MFLX40619-99 MFLX40619-98
Masterflex® Luer Lock Fittings, Stopcock, Avantor®
Valves Ball Valves
Build leakproof fluid handling systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
These one-way stopcocks feature an all-black PVDF construction.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty