Would you like to log in again?
Masterflex® Reducer Fittings, Hose Barb, Straight, Stainless Steel
Supplier: Avantor Fluid Handling
Masterflex®
C12-8SS RC5-3 EA C2-1.5SS C4-3SS RC8-6 EA RC6-3 EA C8-4SS
MFLX31208-14EA 73.1 USD InStock
MFLX31208-14 MFLX31208-13 MFLX30620-12 MFLX31208-15 MFLX30620-11 MFLX30620-10 MFLX30620-09
Masterflex® Reducer Fittings, Hose Barb, Straight, Stainless Steel
Fittings and Connectors
Ideal for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. 316 stainless steel material features very good chemical resistance with a wide temperature range. Sterilize via ethylene oxide, autoclave, or gamma irradiation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty