Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Tee, Stainless Steel, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
T0-12SS T0-8SS T0-5SS T0-2SS T1 EA T0-4SS T0-10SS T0-6SS T6 EA T8 EA T3 EA T5 EA T0-3SS
MFLX30620-29EA 106 USD InStock
MFLX30620-29 MFLX30620-28 MFLX30620-27 MFLX30620-26 MFLX30620-25 MFLX31208-35 MFLX31208-37 MFLX30620-32 MFLX30620-31 MFLX30620-30 MFLX31208-32 MFLX31208-31 MFLX31208-34
Masterflex® Union Fittings, Hose Barb, Tee, Stainless Steel, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty