Would you like to log in again?
Fittings, Compression Nut Assembly with Ferrule
Supplier: MIDWEST VALVE & FITTING INC
IBD4FS+N P-PG-10 K-0-2 K-PG-8 ISSD8FS+N ISSD2FS+N K-PG-4 IBD1FS+N IBD6FS+N IBD2FS+N P-0-2 ISSD1FS+N ISSD6FS+N ISSD4FS+N
MFLX06380-60EA 8.3 USD InStock
MFLX06380-60 MFLX31212-70 MFLX31212-71 MFLX31212-65 MFLX31212-66 MFLX31212-67 MFLX06380-65 MFLX31212-68 MFLX06380-42 MFLX31212-63 MFLX06380-61 MFLX31212-64 MFLX31212-69 MFLX06380-05
Fittings, Compression Nut Assembly with Ferrule
Fittings and Connectors
Can handle pressures up to 225 psi.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty