Would you like to log in again?
CPC® High-Flow Quick-Disconnect Fittings, Bulkhead Panel Mount Hose Barb Bodies
Supplier: CPC (Colder Products Company)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HFCD16612 (10PK) HFC16835 HFC16812 (10PK) HFCD161212 HFCD16835 HFCD16835 10PK HFC161212 (1/PK) HFCD161235 HFC16812 (1/PK) HFCD161235 HFCD16635 (10PK) HFCD16812 (1/PK) HFCD16812 (10PK) HFC16612 HFC161012 HFCD16635 (1/PK) HFCD16612 (1/PK) HFCD161235 10PK HFC161212 (10PK) HFCD161212 HFC161012 10PK
MFLX31308-65BLKEA 257 USD InStock
MFLX31308-65BLK MFLX31303-26 MFLX31308-38 MFLX31303-20 MFLX31308-38BLK MFLX31303-21 MFLX31308-69 MFLX31308-36 MFLX31307-20BLK MFLX31308-67BLK MFLX31308-37 MFLX31308-67 MFLX31307-20 MFLX31308-65 MFLX31307-23 MFLX31308-36BLK MFLX31303-26BLK MFLX31303-28BLK MFLX31303-20BLK MFLX31303-21BLK MFLX31303-28
CPC® High-Flow Quick-Disconnect Fittings, Bulkhead Panel Mount Hose Barb Bodies
Fittings and Connectors
No more leaks—valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty