Would you like to log in again?
CPC® Quick-Disconnect Fittings, Push-to-Connect Inserts
Supplier: CPC (Colder Products Company)
PLC29004 (25PK) PLCD41004 PLC29006 PLC29004 (1/PK) PLCD29004 (1/PK) PLCD29006 (25PK) PLCD41004 25PK PLCD29006 (1/PK) PLCD29004 (25PK)
MFLX31304-32BLKEA 296 USD InStock
MFLX31304-32BLK MFLX31304-20BLK MFLX31304-32 MFLX31304-22 MFLX31308-58 MFLX31308-58BLK MFLX31304-30 MFLX31304-20 MFLX31304-30BLK
CPC® Quick-Disconnect Fittings, Push-to-Connect Inserts
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty