Would you like to log in again?
CPC® Metal Quick-Disconnect Fittings, Compression Inserts
Supplier: CPC (Colder Products Company)
MCD2004 (1/PK) MC2004 (1/PK) MCD2006 10PK LCD20006 MCD2004 (10PK) MCD20025 LCD20004 10PK MC20025 LC20004 MC2004 (10PK) LCD20004 LCD20006 10PK MCD20025 10PK MCD2006 MC20025 10PK LC20006
MFLX31309-19EA 280 USD InStock
MFLX31309-19 MFLX31308-85BLK MFLX31309-11BLK MFLX31404-69 MFLX31309-11 MFLX31403-35 MFLX31309-19BLK MFLX31404-49 MFLX31308-85 MFLX31403-36 MFLX31309-21 MFLX31308-84 MFLX31309-21BLK MFLX31308-84BLK MFLX31404-49BLK MFLX31404-69BLK
CPC® Metal Quick-Disconnect Fittings, Compression Inserts
Fittings and Connectors
Convenience of a quick-disconnect in pressure and vacuum applications.
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180 °F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty