Would you like to log in again?
CPC® Plastic Quick-Disconnect Fittings, Hose Barb Bodies
Supplier: CPC (Colder Products Company)
MPX17839 (5PK) MPCK17006T39 (5PK) MPX17803 (5PK) MPC17004T03 (5PK) MPC17006T39 (5PK) APC17004 (250PK) MPC17004T (250PK) APC17006 (5PK) MPC17004T03 (250PK) MPC17004T39 (250PK) MPXK17839 (250PK) MPCK17004T03 (250PK) APC17006 (25PK) APC17004 (5PK) MPCK17004T03 (5PK) MPXK17803 (250PK) MPCK17006T39 (250PK) MPCK17006T03 (250PK) MPCK17004T39 (5PK) MPXK17803 (5PK) MPXK17839 (5PK) MPX17839 (250PK) MPC17006T MPX17803 (250PK) MPC17006T39 (250PK) MPCK17006T03 (5PK) MPCK17004T39 (250PK) MPC17004T39 (5PK) MPC17006T03 (250PK) MPC17006T03 (5PK)
MFLX31311-18BLKEA 1660 USD InStock
MFLX31311-18BLK MFLX31311-16BLK MFLX31311-26 MFLX31311-28 MFLX31311-26BLK MFLX31311-27 MFLX06360-94 MFLX31311-22BLK MFLX06360-95 MFLX31311-11 MFLX31311-27BLK MFLX31311-13 MFLX06360-94BLK MFLX31311-12 MFLX31311-13BLK MFLX31311-11BLK MFLX31311-17BLK MFLX31311-23BLK MFLX31311-17 MFLX31311-16 MFLX31311-21BLK MFLX31311-18 MFLX31311-22 MFLX31311-28BLK MFLX31311-21 MFLX31311-02 MFLX31311-23 MFLX31311-01 MFLX06360-95BLK MFLX31311-12BLK
CPC® Plastic Quick-Disconnect Fittings, Hose Barb Bodies
Fittings and Connectors
Fittings connect in a snap.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Acetal and ABS fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty