Would you like to log in again?
PFA Adapter Fittings, Compression to Male Threaded, Straight
Supplier: ANDON SPECIALTIES INC.
C10M6N C4M-4N C4M-2N C6M4N
MFLX31321-45EA 77.4 USD InStock
MFLX31321-45 MFLX31321-43 MFLX31321-63 MFLX31321-62
PFA Adapter Fittings, Compression to Male Threaded, Straight
Fittings and Connectors
Superior chemical resistance with durable PFA.
  • Excellent choice to connect rigid tubing systems
  • Ideal for general-purpose and single-use applications
These PFA compression to male thread adapter fittings are ideal for use in corrosive environments and chemical applications. Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty