Would you like to log in again?
Stainless Steel Male Threaded Elbow to Push-to-Connect Adapters
Supplier: WILLIAM P NUGENT COMPANY INC
Masterflex®
PC69SS-86-316 PC69SS-66-316 PC69SS-44-316 PC69SS-88-316 PC69SS-64-316 PC69SS-42-316
MFLX31370-09EA 75.3 USD InStock
MFLX31370-09 MFLX31370-08 MFLX31370-07 MFLX31370-06 MFLX31370-11 MFLX31370-10
Stainless Steel Male Threaded Elbow to Push-to-Connect Adapters
Fittings and Connectors
The ease of push-to-connect and the durability of 316 stainless steel.
  • Constructed of durable stainless steel
The durable stainless steel construction makes these fittings ideal for harsh environments that demand chemically resistant fluid lines. Whether required for a process area with frequent detergent washdowns or exposure to highly corrosive chemicals, these fittings ensure uninterrupted air or fluid lines. Matching material of construction to your stainless piping will reduce the effects of thermal expansion differentials and minimize line leakage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty