Would you like to log in again?
CPC® Metal Quick-Disconnect Fittings, Hose Barb Body
Supplier: CPC (Colder Products Company)
LCD17004 (1/PK) LCD17006 (10PK) MCD1702 MC1702 MCD1702 10PK LCD17006 (1/PK) LC17005 MC1703 MCD1704 LC17006 10PK LC17006 LCD17005 LCD17004 (10PK) LC17004 LCD17005 10PK MC1704
MFLX31309-06EA 341 USD InStock
MFLX31309-06 MFLX31404-10 MFLX31404-32 MFLX31403-20 MFLX31309-05 MFLX31404-12 MFLX31403-22 MFLX31403-25 MFLX31308-83 MFLX31403-22BLK MFLX31308-81 MFLX31308-82 MFLX31308-81BLK MFLX31309-06BLK MFLX31308-83BLK MFLX31403-20BLK
CPC® Metal Quick-Disconnect Fittings, Hose Barb Body
Fittings and Connectors
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180 °F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty