Would you like to log in again?
CPC® Metal Quick-Disconnect Fittings, NPT (F) Threaded Inserts
Supplier: CPC (Colder Products Company)
LCD26004 (1/PK) LC26004 (1/PK) MC2602 (10PK) LC26004 (10PK) MC2602 (1/PK) MC2604 MCD2602 (10PK) MCD2602 (1/PK) LCD26004 (10PK)
MFLX31404-42BLKEA 81.7 USD InStock
MFLX31404-42BLK MFLX31403-57BLK MFLX31404-62 MFLX31403-52 MFLX31404-43 MFLX31404-42 MFLX31403-52BLK MFLX31403-57 MFLX31404-62BLK
CPC® Metal Quick-Disconnect Fittings, NPT (F) Threaded Inserts
Fittings and Connectors
Convenience of a quick-disconnect in pressure and vacuum applications.
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180 °F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty