Would you like to log in again?
CPC® ChemQuik® Quick-Disconnect Fittings, In-Line Coupling Bodies
Supplier: CPC (Colder Products Company)
CQHD06100104 CQHD06100106 CQHD06100108 CQHD06100104 CQHD06100206
MFLX31603-15EA 158.7 USD InStock
MFLX31603-15 MFLX31603-15BLK MFLX31603-01 MFLX31603-02 MFLX31603-00
CPC® ChemQuik® Quick-Disconnect Fittings, In-Line Coupling Bodies
Fittings and Connectors
All plastic construction for high-purity use.
  • 100% metal-free couplings are ideal for ultrapure fluids, deionized water, and aggressive chemical applications
These ChemQuik® quick-disconnect couplings provide easy single-handed disconnection. These couplings feature valves to provide flow shut-off. The all plastic construction and lubricant-free design enables use in the most demanding applications.
For a complete coupling, order one body and one insert of the same size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty