Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CPC® Non-Spill Quick-Disconnect Fittings, Bodies and Inserts

Supplier: CPC (Colder Products Company)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

NS6D10008 (1/PK) NS4D24004BSPT NS6D42008 NS4D22002 1879800 NS4D24004 NS6D40008 NS6D17006 (1/PK) NS4D22006 (1/PK) NS4D10004 NS4D42004 NS4D42006 NS6D17008 (1/PK) NS4D40006 NS6D22006 NS4D24004BSPT (1/PK) NS4D17004 (1/PK) 1884300 NS4D17006 (1/PK) NS4D22004 NS4D10004BSPT NS4D17002 NS6D22008 (1/PK) NS6D24008 (1/PK)
MFLX31603-36EA 27 USD InStock
MFLX31603-36 MFLX31603-26 MFLX31603-23 MFLX31603-34 MFLX31603-35 MFLX31603-24 MFLX31603-18 MFLX31603-29 MFLX31603-19 MFLX31605-08 MFLX31603-27 MFLX31309-49 MFLX31309-46 MFLX31309-47 MFLX31603-40 MFLX31309-44 MFLX31605-13 MFLX31603-32 MFLX31603-22 MFLX31603-44 MFLX31603-33 MFLX31309-43 MFLX31603-31 MFLX31309-41
CPC® Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Fittings and Connectors
Stop leaks in your line—spillage as low as 0.10 ml.

  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety
  • Easy one-hand operation

These CPC® non-spill quick-disconnect fittings virtually eliminate any loss of product with this leak-free design. Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance. This non-spill series ensures minimal leaks and air inclusions in your process line—protects process purity and minimizes loss due to accidental disconnects. The NS4 and NS6 series non-spill couplings confirm secure engagement with an audible click.

The NS4 series offers a spillage rate of <0.1 ml. The NS6 series offers a spillage rate of <0.3 ml.

Choose one body and one insert, each from the same coupling series (NS4 or NS6) for a fitting that will work in multiple applications.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty