Would you like to log in again?
CPC® DrumQuik® PRO Drum Dispensing System Components
Supplier: CPC (Colder Products Company)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2403000 2402900 2879800 (1/PK) 2402700 DQPRO2056X4 DQPRODT0985 (25PK) DQPRO120208 (5PK) DQPRODT1070 DQPRO120208 (1/PK) 2402800 DQPRODT0985 (1/PK) 2403100 DQPRO120712 (1/PK) 2007900 DQPRODT0410 DQPRO120712 (5PK) DQPRO202BUT (25PK) DQPRO202BUT (1/PK) 2403100 2879800 (25PK)
MFLX31608-35EA 7.8 USD InStock
MFLX31608-35 MFLX31608-25 MFLX31309-66 MFLX31608-26 MFLX31608-37 MFLX31608-38 MFLX31309-67 MFLX31608-31 MFLX31608-20 MFLX31608-25BLK MFLX31309-65 MFLX31608-21 MFLX31608-20BLK MFLX31608-22 MFLX31608-34 MFLX31608-23 MFLX31309-63 MFLX31608-21BLK MFLX31608-22BLK MFLX31608-38BLK
CPC® DrumQuik® PRO Drum Dispensing System Components
Fittings and Connectors
Closed-system design ensures the safety of your environment and employees—at all phases of chemical handling.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Indivdual components to keep your dispensing system working smoothly—or build your own
  • Easy-to-use, economical solution for chemical dispensing
  • Durable materials withstand harsh operating conditions
  • Ideal for 5-gallon jerry cans, 55-gallon drums, and IBCs
The CPC® DrumQuik® PRO drum dispensing system offers a simplified solution to your chemical dispensing needs. This system maintains a closed system, protecting your facility, your operating environment, and your employees from the potential dangers of chemical drips, spills and escaping fumes.
The simple connections, rugged construction, and chemical-resistant materials handle a wide variety of applications—including acids and caustics, agricultural liquids, cleaning agents and sterilant, paints and coatings, and food oils and syrups.
Build your own system with these individual components. Required system components are a coupler body, drum insert, dip tube, and liquid port coupler. Optional accessories include a vent check valve and drum insert plug.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty