Would you like to log in again?
Masterflex® Adapter Fittings, Sanitary Clamp to Hose Barb, Straight, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
TAD100L-02H TAD200-32H KAD100L-02H KAD200-16H KAD075-08H KAD200-20H KAD075-04H KAD075-06H TAD075-02H KAD100L-12H TAD200-16H TAD100L-04H TAD150-24H KAD100L-16H KAD100L-06H KAD150-16H TAD075-12H KAD100L-08H KAD075-02H KAD100L-06H KAD075-02H KAD100L-04H KAD100L-12H KAD100L-04H KAD150-16H TAD100L-16H KAD200-16H KAD075-12H TAD100L-12H TAD075-08H KAD075-06H TAD100L-08H TAD075-04H TAD150-16H KAD200-24H KAD075-08H KAD200-32H KAD075-10H KAD100L-08H TAD075-06H KAD075-04H KAD200-32H KAD100L-02H TAD100L-06H KAD075-12H
MFLX31808-31EA 289 USD InStock
MFLX31808-31 MFLX40619-20 MFLX31808-32 MFLX40619-21 MFLX31808-30 MFLX31808-50 MFLX40619-28 MFLX31805-24 MFLX40619-29 MFLX40619-26 MFLX31805-26 MFLX31805-47 MFLX40619-27 MFLX31805-25 MFLX40619-24 MFLX40619-25 MFLX31805-41 MFLX40619-22 MFLX31808-33 MFLX40619-23 MFLX31808-34 MFLX31805-27 MFLX31805-29 MFLX31805-50 MFLX31808-41 MFLX40619-30 MFLX40619-17 MFLX31805-35 MFLX31805-34 MFLX40619-18 MFLX31808-29 MFLX40619-15 MFLX31808-26 MFLX40619-16 MFLX31808-27 MFLX31805-31 MFLX31808-24 MFLX31805-30 MFLX31808-47 MFLX40619-14 MFLX31808-25 MFLX31805-33 MFLX31805-32 MFLX49007-09 MFLX40619-19
Masterflex® Adapter Fittings, Sanitary Clamp to Hose Barb, Straight, Avantor®
Fittings and Connectors
Maintain sanitary conditions while quickly connecting to tubing.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
Allowing for easy assembly and disassembly of sanitary set-ups, these adapters are suitable for use connecting a Tri-Clamp® fitting to flexible tubing with the hose barb adapter.
Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving.
Clamps and gaskets required for a complete system are sold separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty