Would you like to log in again?
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Water, 150-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
PMR1-010741 PMR1-010805 PMR1-010721 PMR1-010788 PMR1-010640 PMR1-010720 P04/1-N102-11G PMR1-010809 PMR1-010786 PMR1-010764 PMR1-010574 PMR1-010613 PMR1-010810 PMR1-010807 PMR1-010745 PMR1-010744 P04/1-044-15ST PMR1-010806 P04/1-N034-10ST PMR1-010639 PMR1-010698 PMR1-010726 PMR1-010787 PMR1-010763 PMR1-010723 PMR1-010808 PMR1-010701 PMR1-011958 PMR1-010743 PMR1-010703 PMR1-010600 PMR1-010719 PMR1-010742 PMR1-010722 PMR1-010765
MFLX32003-38EA 218 USD InStock
MFLX32003-38 MFLX32001-36 MFLX32003-36 MFLX32001-34 MFLX32005-38 MFLX32000-28 MFLX32002-28 MFLX32001-38 MFLX32032-40 MFLX32004-34 MFLX32002-30 MFLX32000-34 MFLX32002-36 MFLX32002-34 MFLX32004-36 MFLX32000-38 MFLX32001-28 MFLX32005-28 MFLX32004-38 MFLX32002-38 MFLX32003-28 MFLX32030-40 MFLX32033-42 MFLX32035-42 MFLX32033-40 MFLX32035-40 MFLX32003-30 MFLX32005-34 MFLX32005-36 MFLX32003-34 MFLX32001-32 MFLX32005-30 MFLX32003-32 MFLX32001-30 MFLX32005-32
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Water, 150-mm Scale, Avantor®
Flow Meters
No more eye strain—front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
Delivery information: Includes 150-mesh inlet screen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty