Would you like to log in again?
Masterflex® Direct-Read Variable-Area Flowmeters with Valve, Panel-Mount, 65-mm, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
PMR1-010929 PMR1-010889 PMR1-010918 PMR1-010753 PMR1-010910 PMR1-010903 PMR1-010917 PMR1-010880 PMR1-010915 PMR1-010902 PMR1-010876 PMR1-010931 PMR1-010911 PMR1-010921 PMR1-010922 PMR1-010926 PMR1-010899 P24/6-N064-05G PMR1-010919 PMR1-010888 PMR1-010895 PMR1-010927 PMR1-010887 PMR1-010877 PMR1-010896 PMR1-010928 PMR1-010882 PMR1-010884 PMR1-010916 GF-8341-2009 PMR1-010873 PMR1-010920 PMR1-010795 PMR1-010901 PMR1-010900 PMR1-010906 PMR1-010881 PMR1-010891 PMR1-010885 PMR1-010874 PMR1-010875 PMR1-010890 PMR1-010894 PMR1-010892 PMR1-010908 PMR1-010932 PMR1-010893 PMR1-010898 PMR1-010897 PMR1-010879 PMR1-010923 PMR1-010878 PMR1-010933 GF-8341-2114 PMR1-010883 PMR1-010924 PMR1-010773
MFLX32004-09EA 189 USD InStock
MFLX32004-09 MFLX32014-11 MFLX32015-23 MFLX32015-24 MFLX32015-21 MFLX32013-35 MFLX32015-16 MFLX32014-05 MFLX32015-17 MFLX32015-15 MFLX32014-03 MFLX32014-09 MFLX32015-19 MFLX32014-07 MFLX32003-09 MFLX32015-31 MFLX32015-35 MFLX32013-11 MFLX32014-21 MFLX32015-33 MFLX32014-15 MFLX32014-16 MFLX32013-01 MFLX32014-13 MFLX32013-03 MFLX32013-05 MFLX32014-17 MFLX32015-29 MFLX32013-07 MFLX32013-09 MFLX32015-01 MFLX32013-21 MFLX32013-13 MFLX32013-15 MFLX32014-24 MFLX32013-17 MFLX32013-16 MFLX32014-28 MFLX32013-19 MFLX32014-29 MFLX32013-18 MFLX32005-09 MFLX32013-31 MFLX32015-13 MFLX32014-01 MFLX32013-33 MFLX32015-11 MFLX32015-05 MFLX32013-24 MFLX32015-03 MFLX32013-28 MFLX32015-09 MFLX32013-27 MFLX32015-07 MFLX32013-29
Masterflex® Direct-Read Variable-Area Flowmeters with Valve, Panel-Mount, 65-mm, Avantor®
Flow Meters
No more eye strain - front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
Delivery information: Includes 150-mesh screen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty