Would you like to log in again?
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Air, 150-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
PMR1-010584 PMR1-010690 PMR1-010688 P13/1-112-19C PMR1-010734 PMR1-010712 PMR1-013771 GF-1300 PMR1-010727 PMR1-010706 PMR1-010596 PMR1-010597 PMR1-010735 PMR1-010710 PMR1-010714 PMR1-013703 P03/1-112-19C PMR1-010570 PMR1-010594 PMR1-010567 PMR1-010568 PMR1-010687 PMR1-010696 P23/1-112-19C PMR1-010579 PMR1-010592 PMR1-016668 PMR1-010730 PMR1-010729 PMR1-010713 PMR1-010708 PMR1-010689 PMR1-010571 PMR1-010709 PMR1-010692 PMR1-010715 PMR1-010731 PMR1-010718 PMR1-010593 PMR1-016607 PMR1-010733 PMR1-010694 PMR1-010572 GF-6500
MFLX32002-04EA 328 USD InStock
MFLX32002-04 MFLX32000-02 MFLX32001-14 MFLX32000-22 MFLX32001-12 MFLX32000-00 MFLX32030-15 MFLX32002-08 MFLX32000-06 MFLX32001-18 MFLX32030-16 MFLX32002-06 MFLX32000-04 MFLX32001-16 MFLX32032-14 MFLX32032-16 MFLX32000-08 MFLX32032-15 MFLX32030-14 MFLX32032-10 MFLX32031-00 MFLX32030-10 MFLX32002-10 MFLX32030-08 MFLX32000-12 MFLX32001-24 MFLX32002-14 MFLX32030-09 MFLX32001-00 MFLX32000-10 MFLX32002-12 MFLX32001-06 MFLX32032-09 MFLX32000-14 MFLX32001-04 MFLX32032-08 MFLX32030-00 MFLX32031-15 MFLX32001-08 MFLX32000-18 MFLX32031-10 MFLX32032-00 MFLX32002-00 MFLX32001-10
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Air, 150-mm Scale, Avantor®
Flow Meters
No more eye strain—front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
Delivery information: Includes 150-mesh inlet screen.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty