Would you like to log in again?
Masterflex® Direct-Read Variable-Area Flowmeters with Valve for Air, 150-mm, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
PMR1-010762 PMR1-011228 PMR1-010791 PMR1-010754 GF-8521-2417 PMR1-010758 PMR1-010605 PMR1-010623 PMR1-010796 PMR1-010610 PMR1-010790 PMR1-010792 PMR1-010782 PMR1-010608 PMR1-010794 PMR1-010793 PMR1-010769 PMR1-010759 PMR1-010618 PMR1-010755 PMR1-010800 PMR1-010761 PMR1-010634 PMR1-016716 PMR1-010756 PMR1-010775 PMR1-010624 PMR1-010804 PMR1-010637 PMR1-010751 PMR1-010803 PMR1-010774 PMR1-010631 PMR1-010750 PMR1-010772 PMR1-010752 PMR1-010611 PMR1-010620 PMR1-010622 PMR1-010773 PMR1-010801 PMR1-010636 PMR1-010797 PMR1-010606 PMR1-010798 PMR1-010635 PMR1-010777 PMR1-010621 PMR1-010633 PMR1-010748 PMR1-010632 PMR1-010749 PMR1-010757 PMR1-010619 PMR1-010607 GF-8521-2317 P14/1-042-75C
MFLX32004-08EA 244 USD InStock
MFLX32004-08 MFLX32033-09 MFLX32004-04 MFLX32005-18 MFLX32004-06 MFLX32033-08 MFLX32034-15 MFLX32034-16 MFLX32034-14 MFLX32033-00 MFLX32004-10 MFLX32004-12 MFLX32005-24 MFLX32003-00 MFLX32005-20 MFLX32003-04 MFLX32003-02 MFLX32004-14 MFLX32005-26 MFLX32003-08 MFLX32004-16 MFLX32034-08 MFLX32003-06 MFLX32034-09 MFLX32035-16 MFLX32035-14 MFLX32035-15 MFLX32034-00 MFLX32034-10 MFLX32003-12 MFLX32003-10 MFLX32003-16 MFLX32003-14 MFLX32004-26 MFLX32003-18 MFLX32035-09 MFLX32035-08 MFLX32035-10 MFLX32005-00 MFLX32005-02 MFLX32003-20 MFLX32005-08 MFLX32003-26 MFLX32003-24 MFLX32005-04 MFLX32005-06 MFLX32033-16 MFLX32033-14 MFLX32033-15 MFLX32033-10 MFLX32035-00 MFLX32004-00 MFLX32005-12 MFLX32005-14 MFLX32005-10
Masterflex® Direct-Read Variable-Area Flowmeters with Valve for Air, 150-mm, Avantor®
Flow Meters
No more eye strain - front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
Delivery information: Includes 150-mesh inlet screen.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty