Would you like to log in again?
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, 65-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
P21/6-N054-17G P11/6-N032-15ST P01/6-NO14-96G P21/6-N054-17ST P01/6-NO23-92ST P11/6-N023-92ST P01/6-NO22-13ST P21/6-N013-88G P21/6-N014/96ST P21/6-N032-15ST P21/6-N022-13G P11/6-N042-07ST P01/6-NO32-15G P11/6-N042-07G P01/6-NO54-17ST P01/6-N0365ST P21/N032-15G P21/6-052-01ST P01/6-NO42-07ST P01/6-NO12-10G P21/6-N052-01G P11/6-N022-13G P01/6-NO23-92G P01/6-NO54-17G P01/6-N0365G P01/6-NO13-88G P01/6-NO13-88ST P21/6-N023-92ST P21/6-N042-07ST P01/6-N052-01G P11/6-N052-01G P21/6-N042-07G P11/6-N013-88G P01/6-NO42-07G P21/6-N012-10G P21/6-N022-13ST P11/6-N022-13ST P11/6-N0365ST P21/6-N013-88ST P11/6-N012-10G P01/6-NO14-96ST P21/6-N012-10ST P11/6-N012-10ST P01/6-NO32-15ST P11/6-N052-01ST P11/6-N054-17G P01/6-NO22-13G P01/6-NO12-10ST P11/6-N032-15G
MFLX32045-32EA 243 USD InStock
MFLX32045-32 MFLX32044-20 MFLX32044-18 MFLX32044-14 MFLX32044-15 MFLX32045-26 MFLX32044-16 MFLX32046-35 MFLX32044-10 MFLX32044-12 MFLX32046-31 MFLX32046-33 MFLX32045-20 MFLX32044-08 MFLX32044-04 MFLX32045-15 MFLX32045-18 MFLX32044-06 MFLX32045-17 MFLX32045-12 MFLX32044-00 MFLX32046-23 MFLX32045-14 MFLX32044-02 MFLX32045-10 MFLX32046-21 MFLX32045-08 MFLX32046-16 MFLX32045-04 MFLX32046-19 MFLX32045-06 MFLX32046-18 MFLX32044-32 MFLX32046-13 MFLX32045-00 MFLX32044-34 MFLX32046-15 MFLX32045-02 MFLX32046-11 MFLX32044-30 MFLX32046-09 MFLX32044-26 MFLX32046-05 MFLX32044-28 MFLX32046-07 MFLX32044-22 MFLX32046-01 MFLX32044-24 MFLX32046-03
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, 65-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty