Would you like to log in again?
Masterflex® Direct Reading Variable Area Flowmeters for Water, PFA Housing, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
L53/B4 GF-1500 L54/B4 L53/A3 L53/B3 L53/B2 L54/A5 L53/B1 GF-1600 L53/A5 L54/B1 L54/C1 L54/A1 L54/A2
MFLX32050-02EA 498 USD InStock
MFLX32050-02 MFLX32050-14 MFLX32050-04 MFLX32050-16 MFLX32051-16 MFLX32051-08 MFLX32051-18 MFLX32050-08 MFLX32051-00 MFLX32050-10 MFLX32051-10 MFLX32050-00 MFLX32051-02 MFLX32050-12
Masterflex® Direct Reading Variable Area Flowmeters for Water, PFA Housing, Avantor®
Flow Meters
A unique combination of rugged frame and chemically inert wetted materials.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with water for laboratory and industrial applications
  • Rigid aluminum frame is PFA coated to resist ambient corrosives
  • Ideal for applications involving ultrapure water or highly corrosive solvents
These flowmeters are great for use with etchants and high purity water like those used in the semiconductor industry. Each flowmeter is individually tested on a mass spectrometer leak detector and certified to a leak integrity rating of 1x10–7 sccs helium or better. A polycarbonate front shield magnifies readings 16% for reading at greater distances. Scale measures 75-mm on flowmeters rated for 5,000 ml/min or less; 125-mm on flowmeters rated to 13 L/min and above. All models have direct-reading water scales.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty