Would you like to log in again?
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Polycarbonate Shield, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
F6E-F10-01-TF F3C-F05-01-TF F3C-F04-01-TF F3C-F01-01-SA F3E-F11-01-TF F3C-F03-01-TF F3E-F10-01-TF F3D-F07-01-TF F6F-F13-01-TF F6C-F05-01-TF F6D-F08-01-TF F3C-F02-01-TF F3F-F12-01-TF F3D-F06-01-TF F6C-F03-01-TF F6D-F07-01-TF F6D-F09-01-TF F6C-F02-01-TF F6C-F01-01-SA F6D-F06-01-TF
MFLX32056-08EA 319 USD InStock
MFLX32056-08 MFLX32057-16 MFLX32056-02 MFLX32057-18 MFLX32056-12 MFLX32056-00 MFLX32057-08 MFLX32056-22 MFLX32056-18 MFLX32057-12 MFLX32056-06 MFLX32057-24 MFLX32057-02 MFLX32057-14 MFLX32056-04 MFLX32057-04 MFLX32057-10 MFLX32057-00 MFLX32056-10 MFLX32056-20
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Polycarbonate Shield, Avantor®
Flow Meters
Economical design includes safety features for metering the most aggressive fluids.
  • Removable polycarbonate shield provides added safety
  • Individually leak tested assures leak-proof integrity*
  • Sight tube guide rod prevents float oscillation in dynamic systems
Ideal for use in applications requiring the high purity/high temperature suitability that fluoropolymers are capable of. Common applications include those with acids, caustics, solvents, and high purity liquids.
The solid PTFE float is designed to maximize each meter's flow range so that many of the available models provide an outstanding turn-down ratio. Flowmeters have direct-reading water scales.
*Leak integrity rating of 1 x 10-7 sccs Helium or better.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty