Would you like to log in again?
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Dual Scale, Aluminum, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
V24/AP V14/BP V13/C V24/BP V14/CP V24/C V24/E V24/B V23/A
MFLX32061-12EA 704 USD InStock
MFLX32061-12 MFLX32062-12 MFLX32061-11 MFLX32061-00 MFLX32062-32 MFLX32062-30 MFLX32061-14 MFLX32060-02 MFLX32062-14
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Dual Scale, Aluminum, Avantor®
Flow Meters
Interchangeable medias and scales offer flexibility.
  • Select water or air scale in either English or metric units by rotating the flowtube
  • Dual water scale shows flow rate in gallons per minute and liters per minute
  • Dual air scale shows flow rate in standard cubic feet per minute and liters per minute
The heavy-walled flowtube construction and protective metal housing make these dual media/scale flowmeters perfect for busy industrial areas. Choose from models with a valve to monitor/control flow or without a valve to just monitor flow. Flowmeters are also available with brass or 316 stainless steel fittings. All models feature an aluminum frame, borosilicate glass tube, 316 stainless steel float, and Viton® O-ring.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty