Would you like to log in again?
Masterflex® Direct-Reading Variable Area Flowmeters for Water, Brass Housing, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
HC205B-020 HC705B-010 HC705B-020 HC755B-020 HC755B-040 HC205B-010 HC605B-005
MFLX32204-24EA 960 USD InStock
MFLX32204-24 MFLX32204-04 MFLX32204-14 MFLX32204-22 MFLX32204-10 MFLX32204-06 MFLX32204-16
Masterflex® Direct-Reading Variable Area Flowmeters for Water, Brass Housing, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Use for high pressure industrial applications—install in any configuration, even next to a bend in your pipeline. A calibrated retention spring eliminates the need to mount flowmeters vertically or run straight lengths of pipe prior to or after the meter. These rugged flowmeters are relatively insensitive to shock and vibrations—the spring, piston assembly, and internal magnet compensate for rough process environments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty