Would you like to log in again?
Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
MS-VJP-M03-01-ST-N MS-VKP-M13-01-ST-N MS-VJP-M05-01-ST-N MS-VJP-M06-01-ST-N MS-VJP-M04-01-ST-N MS-VJP-M02-01-ST-N MS-VKP-M09-01-ST-N MS-VKP-M07-01-ST-N MS-VJP-M01-01-ST-N MS-VLP-M16-01-ST-N MS-VKP-M08-01-ST-N MS-VKP-M11-01-ST-N MS-VKP-M14-01-ST-N MS-VKP-M12-01-ST-N
MFLX32447-50EA 912 USD InStock
MFLX32447-50 MFLX32447-60 MFLX32447-30 MFLX32447-52 MFLX32447-62 MFLX32447-40 MFLX32447-10 MFLX32447-54 MFLX32447-32 MFLX32447-20 MFLX32447-42 MFLX32447-34 MFLX32447-00 MFLX32447-02
Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Flow Meters
Ideal for the most aggressive process applications.
  • Thick polycarbonate front shield provides protection
  • Unique design facilitates ease of maintenance
  • Easy-to-read direct-reading scale offers both metric and English measurements
  • Available in fluted or plain tapered tubes
Detachable, clear 3/16"-thick polycarbonate front shield provides protection at maximum rated temperature and pressure. Easily read the graduated direct-reading scale. The interchangeable transparent scale plates mount directly on the front of the flowtube. Unique float stops allow easy cleaning-remove and replace the float without disturbing either the flowtube or the shield. Tube sizes 3, 4, and 5 are fluted; tube sizes 6, 8, and 9 are tapered with a stainless steel guide rod.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty