Would you like to log in again?
Masterflex® Direct-Read Variable-Area Flowmeters, Air and Water, Acrylic Housing, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
2510A2A00BNBN-CP 2510A2L19BNBN-CP 2520A4A30BNBN-CP 2510A2A06BNBN-CP 2510A2A08BNBN-CP 2520A4L48BNBN-CP 2510A2A03BNBN-CP 2540A5A52PS-CP 2520A4L45BNBN-CP 2540A5L56PS-CP 2520A4A36BNBN-CP 2520A4A33BNBN-CP 2520A4A34BNBN-CP 2510A2L21BNBN-CP 2540A5L58PS-CP 2510A2A02BNBN-CP 2540A4L57PS-CP 2530A4L66BNBN-CP 2510A2A01BNBN-CP 2530A4L64BNBN-CP 2510A2L28BNBN-CP 2530A4A65BNBN-CP 2540A5A50PS-CP 2530A4A67BNBN-CP 2520A4L46BNBN-CP
MFLX32457-26EA 108 USD InStock
MFLX32457-26 MFLX32458-16 MFLX32457-02 MFLX32457-22 MFLX32457-00 MFLX32458-10 MFLX32462-10 MFLX32462-14 MFLX32458-08 MFLX32458-07 MFLX32457-16 MFLX32458-06 MFLX32457-14 MFLX32458-26 MFLX32458-02 MFLX32457-12 MFLX32458-24 MFLX32457-10 MFLX32458-00 MFLX32458-22 MFLX32462-02 MFLX32462-00 MFLX32462-04 MFLX32458-18 MFLX32457-06
Masterflex® Direct-Read Variable-Area Flowmeters, Air and Water, Acrylic Housing, Avantor®
Flow Meters
Durable single-piece body withstands severe shock and vibration.
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct English-Reading Scales
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately below) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters. Contact our Application Specialists for quotes on acrylic meters with special requirements.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty