Would you like to log in again?
Masterflex® Direct-Read Variable-Area Flowmeters with Valve, English Units, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
2510A2A06BVBN-CP 2530A4A65BVBN-CP 2520A4L46BVBN-CP 2510A2A03BVBN-CP 2520A4L45BVBN-CP 2540A4L58PV-CP 2530A4A67BVBN-CP 2520A4A36BVBN-CP 2530A4L66BVBN-CP 2510A2L22BVBN-CP 2510A2L28BVBN-CP 2520A4L48BVBN-CP 2520A4A33BVBN-CP 2520A4A34BVBN-CP 2510A2A01BVBN-CP 2540A4A51PV-CP 2510A2A15BVBN-CP 2510A2L19BVBN-CP 2510A2A02BVBN-CP 2520A4A31BVBN-CP 2510A2A07BVBN-CP 2540A5A52PV-CP 2540A4L57PV-CP 2510A2L21BVBN-CP 2530A4L64BVBN-CP 2520A4A30BVBN-CP 2510A2A00BVBN-CP 2540A5A50PV-CP
MFLX32461-08EA 225 USD InStock
MFLX32461-08 MFLX32461-06 MFLX32460-16 MFLX32461-07 MFLX32461-00 MFLX32461-22 MFLX32460-10 MFLX32461-26 MFLX32460-14 MFLX32461-24 MFLX32461-02 MFLX32460-12 MFLX32466-62 MFLX32466-64 MFLX32466-60 MFLX32460-08 MFLX32461-18 MFLX32460-06 MFLX32460-22 MFLX32460-00 MFLX32461-12 MFLX32461-10 MFLX32461-16 MFLX32460-04 MFLX32460-02 MFLX32460-24 MFLX32466-50 MFLX32466-52
Masterflex® Direct-Read Variable-Area Flowmeters with Valve, English Units, Avantor®
Flow Meters
Meter with valve provides flow control through a highly durable meter body.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate
  • Use with air or water for laboratory and industrial applications
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately) which allows for mobility from bench to bench.
There are many additional types of acrylic flowmeters not listed here. Please contact your local Avantor office for quotes on acrylic meters with special requirements.
Delivery information: Includes flowmeter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty