Would you like to log in again?
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Polysulfone Housing, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
F-452130LHN F-451001LHN F-44376L-8 F-44500L-8 F-451003LHN F-44750L-12 F-451004LHN F-452080LHN F-452060LHN F-44250L-8 F-451002LHN F-44375L-8
MFLX32470-04EA 183 USD InStock
MFLX32470-04 MFLX32473-02 MFLX32472-01 MFLX32472-00 MFLX32473-01 MFLX32473-04 MFLX32472-03 MFLX32470-00 MFLX32472-02 MFLX32470-01 MFLX32470-02 MFLX32470-03
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Polysulfone Housing, Avantor®
Flow Meters
Get better results than acrylic.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Choose from standard, high-capacity, or ultrapure in-line flowmeters. Standard models are designed for low flow readings—measure flow rates as low as 0.025 GPM. The high-capacity flowmeters measure flow rate as high as 130 GPM!
These large, durable flowmeters have 316 SS internal parts. New ultrapure flowmeters use only plastic wetted parts, making these great for ultrapure water such as deionized water. For high-capacity models with polycarbonate shields around the flowtube, call our Application Specialists and get more information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty