Would you like to log in again?
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Dual Scale, Acrylic, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
F-40377LN-6 F-40375LN-8 F-40750LN-16 F-41000LN-16 F-40375LN-6 F-42040LN F-40500LN-6 F-40376LN-6 F-40250LN-6 F-40750LN-12 F-40376LN-8 F-40250LN-4 F-41000LN-12 F-42050LN F-42025LN F-40377LN-8 F-40500LN-8
MFLX32477-20EA 306 USD InStock
MFLX32477-20 MFLX32477-30 MFLX32477-02 MFLX32477-12 MFLX32477-78 MFLX32477-22 MFLX32477-00 MFLX32477-10 MFLX32477-82 MFLX32477-80 MFLX32477-06 MFLX32477-28 MFLX32477-16 MFLX32477-04 MFLX32477-14 MFLX32477-08 MFLX32477-18
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Dual Scale, Acrylic, Avantor®
Flow Meters
Read measurements in an unobstructed 360° view.
  • Easy-to-read dual scales (GPM/LPM)
  • Extremely durable—machined from high-quality acrylic rod stock
  • One-piece, in-line body occupies less space for a clean installation
These meters are designed for use in more aggressive process environments. In addition to the machined acrylic bodies, the design utilizes stainless steel float guides. The combination is a meter suitable for high-vibration, high-fluctuation process applications. Some versions include polypropylene end fittings reinforced with aluminum stress rings for added strength to the meters’ design.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty