Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hedland High TempFlow Meters for Petroleum and Water

Supplier: Hedland
H601A-010-HT H613S-010-HT H613S-002-HT H601A-005-HT H801A-030-HT H601A-002-HT H801S-150-HT
MFLX32508-93EA 467 USD InStock
MFLX32508-93 MFLX32508-94 MFLX32509-40 MFLX32509-09 MFLX32508-92 MFLX32509-07 MFLX32509-31
Hedland High TempFlow Meters for Petroleum and Water
Flow Meters
Direct read 360° rotatable scale.

  • Easy-to-read linear scale

FEATURES
Direct readings
Install in any position
360° Rotatable Guard/Scale
No flow Straighteners or Special Piping Required
Insensitive to Shock and Vibration
Good Viscosity Stability
Temperature up to 500 °F
Accuracy +/- 2% Full Scale
Repeatability +/-1%
Special Scales Available

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty