Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hedland Flow Meters for Air and Other Compressed Gases

Supplier: Hedland

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

H671A-025 H671S-050 H271A-010 H271A-005 H771A-100 H871S-999 H671A-100 H271A-005-EG H671A-150 H271A-020 H871A-600 H671A-150-EG H271A-030 H871A-800-EG H771A-250 H871A-400 H271S-010 H671S-150-EP H671A-050 H271A-010-EG
MFLX32510-63EA 249 USD InStock
MFLX32510-63 MFLX32511-31 MFLX32510-76 MFLX32510-87 MFLX32511-00 MFLX32510-56 MFLX32510-78 MFLX32510-57 MFLX32511-24 MFLX32510-68 MFLX32510-80 MFLX32510-92 MFLX32510-82 MFLX32510-60 MFLX32206-37 MFLX32510-58 MFLX32511-27 MFLX32511-48 MFLX32511-04 MFLX32510-59
Hedland Flow Meters for Air and Other Compressed Gases
Flow Meters
Direct read 360° rotatable scale.

  • Easy-to-read linear scale

FEATURES
Direct readings
Install in any position
360° Rotatable Guard/Scale
No Flow Straighteners or Special Piping Required
Insensitive to Shock and Vibration
Good Viscosity Stability
Temperature up to 240 °F
Accuracy +/-2% Full Scale
Repeatability +/-1%
Special Scales Available

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty