Would you like to log in again?
Masterflex® Paddle-Wheel In-Line Flowmeters, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
RB-500MI-GPM1 RB-100MI-GPM1 RT-375MI-GPM2 RB-150MI-LPM1 RT-500MI-LPM1 RB-150MI-GPM2 RT-375MI-LPM1 RB-750MI-LPM1 RB-375MI-LPM2 RT-375MI-GPM1 RT-500MI-GPM1 RB-375MI-GPM2 RB-150MI-GPM1 RB-750MI-GPM1 RB-375MI-GPM1 RT-750MI-GPM1 RT-375MI-LPM2 RB-500MI-LPM1 RT-750MI-LPM1 RT-150MI-LPM2 RT-200MI-LPM1 RT-200MI-GPM1 RB-200MI-GPM1 RT-100MI-GPM1 RB-375MI-LPM1
MFLX32556-62EA 880 USD InStock
MFLX32556-62 MFLX32556-44 MFLX32556-00 MFLX32556-22 MFLX32556-20 MFLX32556-04 MFLX32556-48 MFLX32556-02 MFLX32556-24 MFLX32556-18 MFLX32556-16 MFLX32556-38 MFLX32556-50 MFLX32556-10 MFLX32556-32 MFLX32556-54 MFLX32556-52 MFLX32556-30 MFLX32556-36 MFLX32556-58 MFLX32556-14 MFLX32556-12 MFLX32556-34 MFLX32556-08 MFLX32556-06
Masterflex® Paddle-Wheel In-Line Flowmeters, Avantor®
Flow Meters
Accurate measurement with virtually zero pressure drop.
  • Paddle wheel technology is accurate, affordable, and relatively easy to install and maintain
  • Ideally suited to open channels and partially filled pipes containing water
  • Factory calibrated for GPM or LPM
These economical in-line flowmeters are battery powered (two AAA batteries included) and thus perfect for remote locations. Enclosure is rated NEMA 4X and can be used in outdoor locations. Sensor is constructed of corrosion-resistant PVDF; in-line NPT(M) fittings are of PP.
Delivery information: Includes two AAA batteries.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty