Would you like to log in again?
Air/N2 Gas Mass Flowmeters with Metal Bodies
Supplier: Aalborg
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GFMS-010001 GFMS-010018 GFMS-010014 GFMS-010016 GFMS-010004 GFMS-010006 GFMS-010030 GFMS-010027 GFMS-010021 GFMS-010003 GFMS-011161 GFMS-010026 GFMS-010023 GFMS-010013 GFMS-010031 GFMS-010017 GFMS-010019 GFMS-010649 GFMS-010005 GFMS-010803 GFMS-011159 GFMS-010002 GFMS-010015 GFMS-010022 GFMS-010009 GFMS-010025 GFMS-010243 GFMS-010008 GFMS-010024 GFMS-010029 GFMS-010007
MFLX32648-04EA 703 USD InStock
MFLX32648-04 MFLX32648-68 MFLX32648-02 MFLX32648-45 MFLX32648-08 MFLX32648-06 MFLX32648-62 MFLX32648-84 MFLX32648-60 MFLX32648-00 MFLX32648-66 MFLX32648-44 MFLX32648-43 MFLX32648-86 MFLX32648-64 MFLX32648-42 MFLX32659-00 MFLX32659-20 MFLX32648-36 MFLX32648-58 MFLX32648-14 MFLX32648-34 MFLX32648-56 MFLX32648-12 MFLX32648-19 MFLX32648-16 MFLX32648-50 MFLX32648-92 MFLX32648-10 MFLX32648-54 MFLX32659-10
Air/N2 Gas Mass Flowmeters with Metal Bodies
Flow Meters
Precisely measure flow rates and totals.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These flowmeters feature an advanced U-shaped sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity. Output data from the meter can be sent via a 0 to 5 VDC or 4 to 20 mA signal; an analog to RS converter is available for data collection and analysis on your PC.
All meters include a detachable 3 1/2-digit LCD that can be tilted up to 90° for easy reading.
Remotely mount the display up to three feet using extension cable 32662-70. The optional totalizer 32650-22 displays accumulated flow. The meter is protected from polarity reversal or short circuit by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum, brass, and Viton®; stainless steel (SS) models have wetted materials of SS and Viton.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77 °F (15 to 25 °C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77 °F (15 to 25 °C), add ±0.15%/ °C full-scale.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty