Would you like to log in again?
Oxygen (O2) Gas Mass Flow Controllers with Metal bodies
Supplier: Aalborg
GFCS-010020 GFCS-010022 GFCS-010072 GFCS-010069 GFCS-010017 GFCS-010023 GFCS-010018 GFCS-010016 GFCS-010073 GFCS-010420 GFCS-010026 GFCS-010664 GFCS-010021 GFCS-010075 GFCS-010024
MFLX32660-40EA 998 USD InStock
MFLX32660-40 MFLX32661-30 MFLX32660-30 MFLX32661-42 MFLX32660-42 MFLX32661-32 MFLX32660-36 MFLX32661-38 MFLX32660-48 MFLX32660-38 MFLX32660-28 MFLX32660-32 MFLX32660-34 MFLX32660-45 MFLX32661-36
Oxygen (O2) Gas Mass Flow Controllers with Metal bodies
Flow Switches
Precisely control flow rates with built-in valve.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced straight-tube sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established by either a local potentiometer or by a remote 4 to 20 mA or 0 to 5 VDC signal. Output data is sent via a 0 to 5 VDC or 4 to 20 mA signal; an analog-to-RS converter is also available for data collection and analysis on your PC.
All controllers include a detachable 3-1/2-digit LCD that can be tilted up to 90° for easy reading. The display is remote mountable to three feet using extension cable 32662-70. The totalizer option 32650-22 will display total accumulated flow.
The controller is protected from polarity reversal or short circuit by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum, brass, and Viton; stainless steel (SS) models have wetted materials of SS and Viton.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77 °F (15 to 25 °C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77 °F (15 to 25 °C), add ±0.15%/ °C full-scale.
Delivery information: Includes nIST-traceable calibration report supplied by the manufacturer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty