Would you like to log in again?
Air/N2 Gas Mass Flow Controllers with Metal bodies
Supplier: Aalborg
GFCS-010065 GFCS-010009 GFCS-010066 GFCS-010060 GFCS-010010 P03/6-N042-07G GFCS-010012 GFCS-011292 GFCS-010004 P03/6-N042-07ST GFCS-010058 GFCS-010481 GFCS-010064 P03/6-N052-01G GFCS-010013 GFCS-010014 GFCS-010008 GFCS-010059 GFCS-010057 GFCS-010056 GFCS-010173 GFCS-010061 GFCS-010007 GFCS-010493 GFCS-011480 GFCS-010054 GFCS-010062 GFCS-011100
MFLX32661-20EA 1367 USD InStock
MFLX32661-20 MFLX32665-00 MFLX32660-04 MFLX32661-16 MFLX32660-06 MFLX32661-12 MFLX32660-00 MFLX32660-22 MFLX32660-02 MFLX32660-24 MFLX32661-14 MFLX32661-19 MFLX32660-08 MFLX32660-20 MFLX32661-10 MFLX32665-10 MFLX32666-00 MFLX32660-14 MFLX32661-04 MFLX32660-16 MFLX32661-06 MFLX32661-22 MFLX32660-10 MFLX32661-00 MFLX32661-24 MFLX32660-12 MFLX32661-08 MFLX32660-19
Air/N2 Gas Mass Flow Controllers with Metal bodies
Flow Switches
Precisely control flow rates with built-in valve.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced straight-tube sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established by either a local potentiometer or by a remote 4 to 20 mA or 0 to 5 VDC signal. Output data is sent via a 0 to 5 VDC or 4 to 20 mA signal; an analog-to-RS converter is also available for data collection and analysis on your PC.
All controllers include a detachable 3-1/2-digit LCD that can be tilted up to 90° for easy reading. The display is remote mountable to three feet using extension cable 32662-70. The totalizer option 32650-22 will display total accumulated flow.
The controller is protected from polarity reversal or short circuit by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum, brass, and Viton; stainless steel (SS) models have wetted materials of SS and Viton.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77 °F (15 to 25 °C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77 °F (15 to 25 °C), add ±0.15%/ °C full-scale.
Delivery information: Includes nIST-traceable calibration report supplied by the manufacturer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty