Would you like to log in again?
McMillan Flow Economical Modular System Flow Sensors
Supplier: McMillan Flow
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
100-6 101-9 100-7 100-5 100-3 100-11 100-14 101-5 100-10 101-6 100-4 101-7 100-12 FS100-9 100-13 101-8 101-3
MFLX32703-52EA 380 USD InStock
MFLX32703-52 MFLX32703-55 MFLX32703-54 MFLX32703-50 MFLX32700-02 MFLX32700-14 MFLX32700-04 MFLX32700-16 MFLX32700-20 MFLX32703-56 MFLX32700-00 MFLX32700-22 MFLX32703-58 MFLX32700-12 MFLX32700-06 MFLX32700-18 MFLX32700-08
McMillan Flow Economical Modular System Flow Sensors
Flow Meters
Ryton® PPS materials for use in aggressive and non-aggressive gas or liquid systems.
  • A range of available configurations to suit most applications
  • Voltage output to monitor and record flow rates and totals
  • Interface with data acquisition system
These sensors are ideal for low-flow applications involving mildly acidic or slightly corrosive gases and liquids. Economically designed, sensors provide a single 0 to 5 VDC output signal. This single signal is ideal for simple, low-cost flow rate measurement or for integration of the sensor into an existing central control system.
Ryton PPS sensors are ideal for liquids or air. Sensors for liquids can be used with a wide variety of transparent, low-viscosity liquids under 10 cSt. Sensors measure a wide flow range from as low as 20 ml/min to as high as 500 L/min.
The wetted materials are epoxy, glass-filled polyphenylene sulfide (Ryton PPS), glass, stainless steel, sapphire, and Viton.
Note: A power supply is required for these flow sensors—order separately. If using an existing power supply, order cable assembly 32704-52.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty