Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Thermal Mass Flowmeters/Flow Transmitters for Gases, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
50D-12-S6 50K-4 50D-6 50K-5 50-11-S6 50D-10 50K-6 50D-15-S7 50K-2 50D-3 50K-9 50D-11-S6 50D-7 50D-13-S7 50D-9 50K-10 50D-2 50D-5 50K-8 50K-7 50K-3 50-12-S6 50D-4 50D-8
MFLX32707-04EA 495 USD InStock
MFLX32707-04 MFLX32707-26 MFLX32707-36 MFLX32707-14 MFLX32707-02 MFLX32707-24 MFLX32707-34 MFLX32707-12 MFLX32707-00 MFLX32707-22 MFLX32707-10 MFLX32707-32 MFLX32707-20 MFLX32707-30 MFLX32707-08 MFLX32707-28 MFLX32707-06 MFLX32707-16 MFLX32711-40 MFLX32712-52 MFLX32712-40 MFLX32712-44 MFLX32711-36 MFLX32712-36
Masterflex® Thermal Mass Flowmeters/Flow Transmitters for Gases, Avantor®
Flow Meters
Get faster performance with miniature coil design.

  • All meters feature 0 to 5 VDC output signal for data logging or controlling other instruments

These low-cost flowmeters use thermal gas flow sensing technique that results in highly accurate readings and repeatability. All units are calibrated to air/nitrogen—call our Application Specialists for meters calibrated to other gases or gas mixtures. Meters feature a cable hub connector for a quick connection to a power supply and 0 to 5 VDC output cable; order power supplies/cables under accessories.

Flowmeters have low pressure drop across the sensor. The output signal can connect to a remote display, recorder, or any instrument that accepts a 0 to 5 VDC signal. Compact size makes it easy to carry around.

Wetted materials are anodized aluminum, 316 stainless steel, Viton® O-rings, and 316 stainless steel, and acetal fittings. Order rechargeable battery kit under Accessories.

Delivery information: Includes two compression fittings and 36" (0.9 m) cable with bub connector.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty