Would you like to log in again?
Masterflex® Paddle Wheel Flowmeters for Monitoring and Control, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
PWM10P PWE04PELNB2 PWM6T PWE010TTLRB2 PWE06TTLRB2 PWE08TTLRB2 PWM8P PWE06PELNB2 PWE06PELRB2 PWE010PELRB2 PWE010PELNB2 PWE04PELRB2
MFLX32755-36EA 975 USD InStock
MFLX32755-36 MFLX32755-24 MFLX32755-46 MFLX32755-35 MFLX32755-38 MFLX32755-32 MFLX32755-31 MFLX32755-45 MFLX32755-34 MFLX32755-44 MFLX32755-18 MFLX32755-17
Masterflex® Paddle Wheel Flowmeters for Monitoring and Control, Avantor®
Flow Meters
Meters provide visual and remote flow indication for numerous applications.
  • Choose from standard or digital models with display and keypad
  • Manufactured from solid PP or PVDF; choose PVDF for better chemical resistance
  • Jewel bearings allow for very low minimum flow rates
  • Only one moving part provides extended life
Designed for water-based liquids, these IP65-rated flowmeters provide flow measurement to 35 GPM (132.5 LPM) and models with the RTD option measure temperature to 140 °F (60 °C). The RTD is platinum encased in 316 stainless steel and has a 4-wire 12" (30.5 cm) cable for output. Flow indication can be seen via acrylic cover. Mount units horizontally or vertically.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty