Would you like to log in again?
Masterflex® Liquid Flow Switches for Threaded Plastic Piping, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
170231 170234 175117 170233 170232
MFLX32774-62EA 203 USD InStock
MFLX32774-62 MFLX32774-64 MFLX32774-66 MFLX32774-68 MFLX32774-60
Masterflex® Liquid Flow Switches for Threaded Plastic Piping, Avantor®
Flow Switches
Connects quickly to threaded plastic piping.
  • Chemically resistant — wetted materials stainless steel and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • NSF and FDA approved
These normally open switches have 3/4" NPT(F) ports for a quick connection to threaded plastic piping. Polypropylene and stainless steel wetted parts offer excellent chemical resistance. The materials are also NSF approved and FDA compliant; approved for potable water treatment applications such as chlorinators, purifiers, and heaters. Switches include 24" lead wires.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty