Would you like to log in again?
Masterflex® Low Pressure-Drop Liquid Flow Switches, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
129666 129671
MFLX32776-20EA 242 USD InStock
MFLX32776-20 MFLX32776-44
Masterflex® Low Pressure-Drop Liquid Flow Switches, Avantor®
Flow Switches
Minimize flow constriction and achieve low pressure drop.
  • Chemically resistant — wetted materials Ryton® PPS, Viton®, 316 Stainless Steel, and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • Durable switch with 212 °F (100 °C) max temperature and 200 psi at 70 °F max pressure
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 µm filter to protect your switch. Switches include 1/4" male quick connect terminals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty