Would you like to log in again?
Masterflex® Vortex Flowmeters with Temperature for Liquids, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
DN15ADS1/2R15EC15RTD, M12X1 DN10ADS3/8R810EC810RTD, M12X1 DN25ADSG1NR25ERTD, M12X1 DN8ADS1/4R810EC810RTD, M12X1 DN20ADS3/4R20EC20RTD, M12X1
MFLX32825-76EA 314 USD InStock
MFLX32825-76 MFLX32825-74 MFLX32825-72 MFLX32825-70 MFLX32825-68
Masterflex® Vortex Flowmeters with Temperature for Liquids, Avantor®
Flow Meters
Built-in 1000 Ω RTD sensor provides direct temperature measurement of media.
  • Deigned with rugged PPA molded construction for enhanced durability
  • Excellent media compatibility with ETFE, EPDM, and PPD wetted parts
  • Use with low viscosity liquids for laboratory and industrial applications
These economical vortex flowmeters are suitable for water and water glycol-based heat exchange systems. The transmitters work on the vortex shedding flow principle. As flow velocity increases, alternating vortices are formed on each side of the bluff body within the flowtube and travel downstream. The flowmeters utilize piezoelectric sensors embedded in a vane downstream from the bluff to detect the vortices and transfer the sensed vortices into flow velocity. A flow factor is provided to convert frequency to volumetric flow rate for each size of flowmeter. Flowmeters have a PT1000 RTD for temperature measurement of the media
Note: Media with viscosity greater than 2 centipose will require higher flow rates to form measurable vortices within the flowtube.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty