Would you like to log in again?
Masterflex® Gas Mass Flowmeters, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
M-250SLPM-D/5M M-1000SLPM-D/5M M-1SCCM-D/5M M-10SLPM-D/5M M-50SCCM-D/5M M-5SCCM-D/5M M-500SLPM-D/5M M-1SLPM-D/5M M-500SCCM-D/5M M-100SLPM-D/5M M-200SCCM-D/5M M-100SCCM-D/5M M-10SCCM-D/5M M-50SLPM-D/5M M-5SLPM-D/5M
MFLX32908-81EA 2960 USD InStock
MFLX32908-81 MFLX32908-61 MFLX32908-71 MFLX32908-63 MFLX32908-51 MFLX32908-73 MFLX32908-53 MFLX32908-75 MFLX32908-67 MFLX32908-77 MFLX32908-55 MFLX32908-69 MFLX32908-57 MFLX32908-79 MFLX32908-59
Masterflex® Gas Mass Flowmeters, Avantor®
Flow Meters
Measure 130 standard gases—user selectable from display.
  • Accuracy of ±0.8% of reading ±0.2% full-scale; repeatability of ±0.2%
  • 100 to 1 turndown with ranges of 1 SCCM full-scale up to 1000 SLPM
  • Models with 4 to 20 mA output or totalizer are available
These meters measure flow via pressure drop across a laminar flow element (LFE). Because the flow element makes the flow stream laminar, placement in the process does not require straight pipe runs upstream or downstream of the meter, greatly simplifying installation. As compared to thermal mass technologies, the LFE design provides an ultrafast response within 10 milliseconds and offers “instant on” with no warm-up time.
An integrated keypad around the display is all that is required to program the unit for service. The 0 to 5 VDC output allows transmission of the flow value to a remote display, recorder, or controller regulating a valve or pump.
For portable flow metering applications, order the battery pack (sold separately).
Delivery information: Includes 120/230 VAC power adapter with communications cable and NIST-traceable calibration report supplied by the manufacturer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty