Would you like to log in again?
Masterflex® Ismatec® Microbore Tubing Straight Connectors, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
34002-12 34002-09 34002-22 34002-10 34002-18 34002-26 34002-24 34002-14
MFLX34002-09EA 108 USD InStock
MFLX34002-09 MFLX34002-18 MFLX34002-26 MFLX34002-14 MFLX34002-24 MFLX34002-12 MFLX34002-22 MFLX34002-10
Masterflex® Ismatec® Microbore Tubing Straight Connectors, Avantor®
Fittings and Connectors
Connect even the smallest tubing with a leak-free seal from vacuum to pressure.
  • Multiple styles to fit your needs
  • Stainless steel fitting sized to match the Ismatec® sizes - from 09 to 26
Delivery information: Each kit includes 2 fittings.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty